3-Chloro-6-methoxy-2,4-dimethylbenzaldehyde
Supplier: Matrix Scientific
053609-500MG
053609-2.5G
101904-322EA
166.15
USD
101904-322
101904-324
3-Chloro-6-methoxy-2,4-dimethylbenzaldehyde
3-Chloro-6-methoxy-2,4-dimethylbenzaldehyde
Hazard: Irritant
Formula:
C₁₀H₁₁ClO₂ Melting Pt: 106…107 °C |
MDL Number:
MFCD06246298 CAS Number: 88174-46-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...