2-(Cyclopropylmethoxy)-5-fluorobenzaldehyde
Supplier: Matrix Scientific
053608-500MG
053608-2.5G
101904-318EA
364.62
USD
101904-318
101904-320
2-(Cyclopropylmethoxy)-5-fluorobenzaldehyde
2-(Cyclopropylmethoxy)-5-fluorobenzaldehyde
Hazard: Irritant
Formula:
C₁₁H₁₁FO₂ |
MDL Number:
MFCD06246758 CAS Number: 883525-60-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...