5-Fluoro-2-propoxybenzaldehyde

Supplier: Matrix Scientific

053607-500MG 053607-2.5G
101904-314EA 461.54 USD
101904-314 101904-316
5-Fluoro-2-propoxybenzaldehyde
5-Fluoro-2-propoxybenzaldehyde

Hazard: Irritant

Formula: C₁₀H₁₁FO₂
MDL Number: MFCD06247718
CAS Number: 19415-54-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR