2-Ethoxy-5-fluorobenzaldehyde
Supplier: Matrix Scientific
Synonyms:
2-Ethoxy-5-fluoro-benzaldehyde
053606-500MG
053606-2.5G
101904-310EA
384.62
USD
101904-310
101904-312
2-Ethoxy-5-fluorobenzaldehyde
2-Ethoxy-5-fluorobenzaldehyde
Hazard: Irritant
Formula:
C₉H₉FO₂ MW: 168.17 g/mol |
MDL Number:
MFCD06247346 CAS Number: 5710-35-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...