4-(Cyclopropylmethoxy)-3-propylbenzaldehyde
Supplier: Matrix Scientific
053605-500MG
053605-2.5G
101904-306EA
364.62
USD
101904-306
101904-308
4-(Cyclopropylmethoxy)-3-propylbenzaldehyde
4-(Cyclopropylmethoxy)-3-propylbenzaldehyde
Hazard: Irritant
Formula:
C₁₄H₁₈O₂ |
MDL Number:
MFCD06246808 CAS Number: 883528-79-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...