4-Propoxy-3-propylbenzaldehyde

Supplier: Matrix Scientific

053604-500MG 053604-2.5G
101904-302EA 364.62 USD
101904-302 101904-304
4-Propoxy-3-propylbenzaldehyde
4-Propoxy-3-propylbenzaldehyde

Hazard: Irritant

Formula: C₁₃H₁₈O₂
MDL Number: MFCD06248744
CAS Number: 883533-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR