4-Ethoxy-3-propylbenzaldehyde

Supplier: Matrix Scientific

053603-500MG 053603-2.5G
101904-298EA 166.15 USD
101904-298 101904-300
4-Ethoxy-3-propylbenzaldehyde
4-Ethoxy-3-propylbenzaldehyde

Hazard: Irritant

Formula: C₁₂H₁₆O₂
MDL Number: MFCD06247413
CAS Number: 883540-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR