4-Methoxy-3-propylbenzaldehyde
Supplier: Matrix Scientific
053602-500MG
053602-2.5G
101904-294EA
461.54
USD
101904-294
101904-296
4-Methoxy-3-propylbenzaldehyde
4-Methoxy-3-propylbenzaldehyde
Hazard: Irritant
Formula:
C₁₁H₁₄O₂ Boiling Pt: 145 °C (1 Torr) |
MDL Number:
MFCD06248103 CAS Number: 104175-21-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...