4-(Cyclopropylmethoxy)-3-ethylbenzaldehyde

Supplier: Matrix Scientific

053601-500MG 053601-2.5G
101904-290EA 364.62 USD
101904-290 101904-292
4-(Cyclopropylmethoxy)-3-ethylbenzaldehyde
4-(Cyclopropylmethoxy)-3-ethylbenzaldehyde

Hazard: Irritant

Formula: C₁₃H₁₆O₂
MDL Number: MFCD06246751
CAS Number: 883525-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR