3-Ethyl-4-methoxybenzaldehyde

Supplier: Matrix Scientific

053598-500MG 053598-2.5G
101904-278EA 364.62 USD
101904-278 101904-280
3-Ethyl-4-methoxybenzaldehyde
3-Ethyl-4-methoxybenzaldehyde

Hazard: Irritant

Formula: C₁₀H₁₂O₂
MDL Number: MFCD06247460
CAS Number: 103323-26-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR