4-(Cyclopropylmethoxy)-3,5-dimethylbenzaldehyde
Supplier: Matrix Scientific
053597-500MG
053597-2.5G
101904-274EA
364.62
USD
101904-274
101904-276
4-(Cyclopropylmethoxy)-3,5-dimethylbenzaldehyde
4-(Cyclopropylmethoxy)-3,5-dimethylbenzaldehyde
Hazard: Irritant
Formula:
C₁₃H₁₆O₂ |
MDL Number:
MFCD06246742 CAS Number: 883524-60-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...