3,5-Dimethyl-4-propoxybenzaldehyde
Supplier: Matrix Scientific
053596-500MG
053596-2.5G
101904-270EA
129.23
USD
101904-270
101904-272
3,5-Dimethyl-4-propoxybenzaldehyde
3,5-Dimethyl-4-propoxybenzaldehyde
Hazard: Irritant
Formula:
C₁₂H₁₆O₂ |
MDL Number:
MFCD06247276 CAS Number: 210057-00-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...