4-Ethoxy-3,5-dimethylbenzaldehyde

Supplier: Matrix Scientific

053595-500MG 053595-2.5G
101904-266EA 253.85 USD
101904-266 101904-268
4-Ethoxy-3,5-dimethylbenzaldehyde
4-Ethoxy-3,5-dimethylbenzaldehyde

Hazard: Irritant

Formula: C₁₁H₁₄O₂
MDL Number: MFCD06247333
CAS Number: 861376-63-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR