2-(Cyclopropylmethoxy)-3,5-dimethylbenzaldehyde
Supplier: Matrix Scientific
Synonyms:
2-(Cyclopropylmethoxy)-3,5-dimethylbenzaldehyde
053594-500MG
053594-2.5G
101904-262EA
364.62
USD
101904-262
101904-264
2-(Cyclopropylmethoxy)-3,5-dimethylbenzaldehyde
2-(Cyclopropylmethoxy)-3,5-dimethylbenzaldehyde
Hazard: Irritant
Formula:
C₁₃H₁₆O₂ MW: 204.27 g/mol |
MDL Number:
MFCD06246737 CAS Number: 883524-34-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...