3,5-Dimethyl-2-propoxybenzaldehyde
Supplier: Matrix Scientific
053593-500MG
053593-2.5G
101904-258EA
384.62
USD
101904-258
101904-260
3,5-Dimethyl-2-propoxybenzaldehyde
3,5-Dimethyl-2-propoxybenzaldehyde
Hazard: Irritant
Formula:
C₁₂H₁₆O₂ |
MDL Number:
MFCD06247275 CAS Number: 883532-80-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...