2-Ethoxy-3,5-dimethylbenzaldehyde

Supplier: Matrix Scientific

053592-500MG 053592-2.5G
101904-254EA 384.62 USD
101904-254 101904-256
2-Ethoxy-3,5-dimethylbenzaldehyde
2-Ethoxy-3,5-dimethylbenzaldehyde

Hazard: Irritant

Formula: C₁₁H₁₄O₂
MDL Number: MFCD06247328
CAS Number: 883536-29-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR