2-Methoxy-3,5-dimethylbenzaldehyde

Supplier: Matrix Scientific

053591-500MG 053591-2.5G
101904-250EA 364.62 USD
101904-250 101904-252
2-Methoxy-3,5-dimethylbenzaldehyde
2-Methoxy-3,5-dimethylbenzaldehyde

Hazard: Irritant

Formula: C₁₀H₁₂O₂
MDL Number: MFCD06247962
CAS Number: 16313-77-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR