4-(Cyclopropylmethoxy)-3-methylbenzaldehyde

Supplier: Matrix Scientific

053590-500MG 053590-2.5G
101904-246EA 461.54 USD
101904-246 101904-248
4-(Cyclopropylmethoxy)-3-methylbenzaldehyde
4-(Cyclopropylmethoxy)-3-methylbenzaldehyde

Hazard: Irritant

Formula: C₁₂H₁₄O₂
MDL Number: MFCD06246776
CAS Number: 820237-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR