3-Methyl-4-propoxybenzaldehyde

Supplier: Matrix Scientific

053589-500MG 053589-2.5G
101904-242EA 364.62 USD
101904-242 101904-244
3-Methyl-4-propoxybenzaldehyde
3-Methyl-4-propoxybenzaldehyde

Hazard: Irritant

Formula: C₁₁H₁₄O₂
MDL Number: MFCD06248397
CAS Number: 820236-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR