4-Ethoxy-3-methylbenzaldehyde

Supplier: Matrix Scientific

053588-500MG 053588-2.5G
101904-238EA 538.46 USD
101904-238 101904-240
4-Ethoxy-3-methylbenzaldehyde
4-Ethoxy-3-methylbenzaldehyde

Hazard: Irritant

Formula: C₁₀H₁₂O₂
MDL Number: MFCD06247361
CAS Number: 56917-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR