3-(sec-Butyl)-4-propoxybenzaldehyde
Supplier: Matrix Scientific
053585-500MG
053585-2.5G
101904-226EA
149.23
USD
101904-226
101904-228
3-(sec-Butyl)-4-propoxybenzaldehyde
3-(sec-Butyl)-4-propoxybenzaldehyde
Hazard: Irritant
Formula:
C₁₄H₂₀O₂ |
MDL Number:
MFCD06246095 CAS Number: 883515-59-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...