3-(sec-Butyl)-4-ethoxybenzaldehyde
Supplier: Matrix Scientific
053584-500MG
053584-2.5G
101904-222EA
364.62
USD
101904-222
101904-224
3-(sec-Butyl)-4-ethoxybenzaldehyde
3-(sec-Butyl)-4-ethoxybenzaldehyde
Hazard: Irritant
Formula:
C₁₃H₁₈O₂ |
MDL Number:
MFCD06246078 CAS Number: 883514-69-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...