3-(sec-Butyl)-4-methoxybenzaldehyde

Supplier: Matrix Scientific

053583-500MG 053583-2.5G
101904-218EA 538.46 USD
101904-218 101904-220
3-(sec-Butyl)-4-methoxybenzaldehyde
3-(sec-Butyl)-4-methoxybenzaldehyde
Formula: C₁₂H₁₆O₂
MDL Number: MFCD06246084
CAS Number: 883515-13-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR