4-(Cyclopropylmethoxy)-3-isopropylbenzaldehyde

Supplier: Matrix Scientific

053582-500MG 053582-2.5G
101904-214EA 364.62 USD
101904-214 101904-216
4-(Cyclopropylmethoxy)-3-isopropylbenzaldehyde
4-(Cyclopropylmethoxy)-3-isopropylbenzaldehyde

Hazard: Irritant

Formula: C₁₄H₁₈O₂
MDL Number: MFCD06246765
CAS Number: 883525-95-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR