3-Isopropyl-4-propoxybenzaldehyde
Supplier: Matrix Scientific
053581-500MG
053581-2.5G
101904-210EA
364.62
USD
101904-210
101904-212
3-Isopropyl-4-propoxybenzaldehyde
3-Isopropyl-4-propoxybenzaldehyde
Hazard: Irritant
Formula:
C₁₃H₁₈O₂ |
MDL Number:
MFCD06247932 CAS Number: 883538-71-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...