4-Ethoxy-3-isopropylbenzaldehyde

Supplier: Matrix Scientific

053580-500MG 053580-2.5G
101904-206EA 461.54 USD
101904-206 101904-208
4-Ethoxy-3-isopropylbenzaldehyde
4-Ethoxy-3-isopropylbenzaldehyde

Hazard: Irritant

Formula: C₁₂H₁₆O₂
MDL Number: MFCD06247352
CAS Number: 883537-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR