3-Isopropyl-4-methoxybenzaldehyde
Supplier: Matrix Scientific
053579-500MG
053579-2.5G
101904-202EA
292.31
USD
101904-202
101904-204
3-Isopropyl-4-methoxybenzaldehyde
3-Isopropyl-4-methoxybenzaldehyde
Hazard: Irritant
Formula:
C₁₁H₁₄O₂ |
MDL Number:
MFCD06247877 CAS Number: 31825-29-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...