3-(tert-Butyl)-4-(cyclopropylmethoxy)benzaldehyde

Supplier: Matrix Scientific

053578-500MG 053578-2.5G
101904-198EA 364.62 USD
101904-198 101904-200
3-(tert-Butyl)-4-(cyclopropylmethoxy)benzaldehyde
3-(tert-Butyl)-4-(cyclopropylmethoxy)benzaldehyde

Hazard: Irritant

Formula: C₁₅H₂₀O₂
MDL Number: MFCD06246072
CAS Number: 883514-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR