3-(tert-Butyl)-4-propoxybenzaldehyde

Supplier: Matrix Scientific

053577-500MG 053577-2.5G
101904-194EA 364.62 USD
101904-194 101904-196
3-(tert-Butyl)-4-propoxybenzaldehyde
3-(tert-Butyl)-4-propoxybenzaldehyde

Hazard: Irritant

Formula: C₁₄H₂₀O₂
MDL Number: MFCD06246097
CAS Number: 170100-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR