3-(tert-Butyl)-4-ethoxybenzaldehyde

Supplier: Matrix Scientific

053576-500MG 053576-2.5G
101904-190EA 364.62 USD
101904-190 101904-192
3-(tert-Butyl)-4-ethoxybenzaldehyde
3-(tert-Butyl)-4-ethoxybenzaldehyde

Hazard: Irritant

Formula: C₁₃H₁₈O₂
MDL Number: MFCD02380390
CAS Number: 681443-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR