3-(tert-Butyl)-4-methoxybenzaldehyde
Supplier: Matrix Scientific
053575-500MG
053575-2.5G
101904-186EA
364.62
USD
101904-186
101904-188
3-(tert-Butyl)-4-methoxybenzaldehyde
3-(tert-Butyl)-4-methoxybenzaldehyde
Hazard: Irritant
Formula:
C₁₂H₁₆O₂ MW: 192.26 g/mol |
MDL Number:
MFCD01104342 CAS Number: 107430-92-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...