6-Propoxy[1,1'-biphenyl]-3-carbaldehyde

Supplier: Matrix Scientific

053573-500MG 053573-2.5G
101904-178EA 364.62 USD
101904-178 101904-180
6-Propoxy[1,1'-biphenyl]-3-carbaldehyde
6-Propoxy[1,1'-biphenyl]-3-carbaldehyde

Hazard: Irritant

Formula: C₁₆H₁₆O₂
MDL Number: MFCD06248730
CAS Number: 883532-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR