3,5-Dichloro-2-(cyclopropylmethoxy)benzaldehyde
Supplier: Matrix Scientific
053572-500MG
053572-2.5G
101904-174EA
364.62
USD
101904-174
101904-176
3,5-Dichloro-2-(cyclopropylmethoxy)benzaldehyde
3,5-Dichloro-2-(cyclopropylmethoxy)benzaldehyde
Hazard: Irritant
Formula:
C₁₁H₁₀Cl₂O₂ |
MDL Number:
MFCD06246843 CAS Number: 883532-17-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...