3,5-Dichloro-2-propoxybenzaldehyde
Supplier: Matrix Scientific
053571-500MG
053571-2.5G
101904-170EA
364.62
USD
101904-170
101904-172
3,5-Dichloro-2-propoxybenzaldehyde
3,5-Dichloro-2-propoxybenzaldehyde
Hazard: Irritant
Formula:
C₁₀H₁₀Cl₂O₂ |
MDL Number:
MFCD06246855 CAS Number: 883532-89-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...