3-Chloro-2-(cyclopropylmethoxy)-5-fluorobenzaldehyde

Supplier: Matrix Scientific

053569-500MG 053569-2.5G
101904-162EA 364.62 USD
101904-162 101904-164
3-Chloro-2-(cyclopropylmethoxy)-5-fluorobenzaldehyde
3-Chloro-2-(cyclopropylmethoxy)-5-fluorobenzaldehyde

Hazard: Irritant

Formula: C₁₁H₁₀ClFO₂
MDL Number: MFCD06246144
CAS Number: 883519-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR