2-[3-(2-Pyridinyl)-1,2,4-oxadiazol-5-yl]acetic acid

Supplier: Matrix Scientific

053568-500MG 053568-2.5G
101904-158EA 364.62 USD
101904-158 101904-160
2-[3-(2-Pyridinyl)-1,2,4-oxadiazol-5-yl]acetic acid
2-[3-(2-Pyridinyl)-1,2,4-oxadiazol-5-yl]acetic acid

Hazard: Irritant

Formula: C₉H₇N₃O₃
MDL Number: MFCD06808696
CAS Number: 761390-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR