2-[3-(3-Pyridinyl)-1,2,4-oxadiazol-5-yl]acetic acid

Supplier: Matrix Scientific

053567-500MG 053567-2.5G
101904-154EA 364.62 USD
101904-154 101904-156
2-[3-(3-Pyridinyl)-1,2,4-oxadiazol-5-yl]acetic acid
2-[3-(3-Pyridinyl)-1,2,4-oxadiazol-5-yl]acetic acid

Hazard: Irritant

Formula: C₉H₇N₃O₃
MDL Number: MFCD06808701
CAS Number: 874832-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR