2-(3-Cyclopropyl-1,2,4-oxadiazol-5-yl)acetic acid

Supplier: Matrix Scientific

053566-500MG 053566-2.5G
101904-150EA 592.31 USD
101904-150 101904-152
2-(3-Cyclopropyl-1,2,4-oxadiazol-5-yl)acetic acid
2-(3-Cyclopropyl-1,2,4-oxadiazol-5-yl)acetic acid

Hazard: Irritant

Formula: C₇H₈N₂O₃
MDL Number: MFCD13562877
CAS Number: 1220030-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR