3-Chloro-5-fluoro-2-propoxybenzaldehyde
Supplier: Matrix Scientific
053565-500MG
053565-2.5G
101904-146EA
364.62
USD
101904-146
101904-148
3-Chloro-5-fluoro-2-propoxybenzaldehyde
3-Chloro-5-fluoro-2-propoxybenzaldehyde
Hazard: Irritant
Formula:
C₁₀H₁₀ClFO₂ |
MDL Number:
MFCD06246284 CAS Number: 883528-05-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...