3-Chloro-5-fluoro-2-methoxybenzaldehyde

Supplier: Matrix Scientific

053563-500MG 053563-2.5G
101904-138EA 538.46 USD
101904-138 101904-140
3-Chloro-5-fluoro-2-methoxybenzaldehyde
3-Chloro-5-fluoro-2-methoxybenzaldehyde

Hazard: Irritant

Formula: C₈H₆ClFO₂
MDL Number: MFCD06246207
CAS Number: 82129-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR