3-Chloro-4-(cyclopropylmethoxy)benzaldehyde
Supplier: Matrix Scientific
053562-500MG
053562-2.5G
101904-134EA
364.62
USD
101904-134
101904-136
3-Chloro-4-(cyclopropylmethoxy)benzaldehyde
3-Chloro-4-(cyclopropylmethoxy)benzaldehyde
Hazard: Irritant
Formula:
C₁₁H₁₁ClO₂ |
MDL Number:
MFCD06246133 CAS Number: 883519-07-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...