3-Chloro-4-propoxybenzaldehyde
Supplier: Matrix Scientific
053561-500MG
053561-2.5G
101904-130EA
364.62
USD
101904-130
101904-132
3-Chloro-4-propoxybenzaldehyde
3-Chloro-4-propoxybenzaldehyde
Hazard: Irritant
Formula:
C₁₀H₁₁ClO₂ |
MDL Number:
MFCD06246436 CAS Number: 99070-71-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...