3-Chloro-4-ethoxybenzaldehyde

Supplier: Matrix Scientific

053560-500MG 053560-2.5G
101904-126EA 364.62 USD
101904-126 101904-128
3-Chloro-4-ethoxybenzaldehyde
3-Chloro-4-ethoxybenzaldehyde

Hazard: Irritant

Formula: C₉H₉ClO₂
MDL Number: MFCD06246171
CAS Number: 99585-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR