3-Amino-4-chloro-N-(tetrahydro-2H-pyran-4-ylmethyl)benzenesulfonamide
Supplier: Matrix Scientific
053559-500MG
053559-2.5G
101904-122EA
364.62
USD
101904-122
101904-124
3-Amino-4-chloro-N-(tetrahydro-2H-pyran-4-ylmethyl)benzenesulfonamide
3-Amino-4-chloro-N-(tetrahydro-2H-pyran-4-ylmethyl)benzenesulfonamide
Hazard: Irritant
Formula:
C₁₂H₁₇ClN₂O₃S |
MDL Number:
MFCD13562876 CAS Number: 1220019-16-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...