N-(Tetrahydro-2H-pyran-4-ylmethyl)-4,5,6,7-tetra-hydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide HCl

Supplier: Matrix Scientific

053558-500MG 053558-2.5G
101904-118EA 364.62 USD
101904-118 101904-120
N-(Tetrahydro-2H-pyran-4-ylmethyl)-4,5,6,7-tetra-hydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide HCl
N-(Tetrahydro-2H-pyran-4-ylmethyl)-4,5,6,7-tetra-hydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide HCl

Hazard: Irritant

Formula: C₁₃H₂₁ClN₄O₂
MDL Number: MFCD13562875
CAS Number: 1220029-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR