N-(Tetrahydro-2H-pyran-4-ylmethyl)-4-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

053557-500MG 053557-2.5G
101904-114EA 364.62 USD
101904-114 101904-116
N-(Tetrahydro-2H-pyran-4-ylmethyl)-4-piperidinecarboxamide hydrochloride
N-(Tetrahydro-2H-pyran-4-ylmethyl)-4-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₃ClN₂O₂
MDL Number: MFCD13562874
CAS Number: 1220021-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR