N-(Tetrahydro-2H-pyran-4-ylmethyl)-3-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

053556-500MG 053556-2.5G
101904-110EA 364.62 USD
101904-110 101904-112
N-(Tetrahydro-2H-pyran-4-ylmethyl)-3-piperidinecarboxamide hydrochloride
N-(Tetrahydro-2H-pyran-4-ylmethyl)-3-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₃ClN₂O₂
MW: 262.78 g/mol
MDL Number: MFCD13562873
CAS Number: 1220029-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR