N-(Tetrahydro-2H-pyran-4-ylmethyl)-2-piperidinecarboxamide hydrochloride
Supplier: Matrix Scientific
053555-500MG
053555-2.5G
101904-106EA
364.62
USD
101904-106
101904-108
N-(Tetrahydro-2H-pyran-4-ylmethyl)-2-piperidinecarboxamide hydrochloride
N-(Tetrahydro-2H-pyran-4-ylmethyl)-2-piperidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₂H₂₃ClN₂O₂ MW: 262.78 g/mol |
MDL Number:
MFCD13562872 CAS Number: 1236256-57-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...