2-Amino-3-phenyl-N-(tetrahydro-2H-pyran-4-ylmethyl)propanamide hydrochloride
Supplier: Matrix Scientific
053553-500MG
053553-2.5G
101904-098EA
364.62
USD
101904-098
101904-100
2-Amino-3-phenyl-N-(tetrahydro-2H-pyran-4-ylmethyl)propanamide hydrochloride
2-Amino-3-phenyl-N-(tetrahydro-2H-pyran-4-ylmethyl)propanamide hydrochloride
Hazard: Irritant
Formula:
C₁₅H₂₃ClN₂O₂ |
MDL Number:
MFCD13562870 CAS Number: 1236254-74-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...