2-Amino-N-(tetrahydro-2H-pyran-4-ylmethyl)propanamide hydrochloride
Supplier: Matrix Scientific
053548-500MG
053548-2.5G
101904-078EA
364.62
USD
101904-078
101904-080
2-Amino-N-(tetrahydro-2H-pyran-4-ylmethyl)propanamide hydrochloride
2-Amino-N-(tetrahydro-2H-pyran-4-ylmethyl)propanamide hydrochloride
Hazard: Irritant
Formula:
C₉H₁₉ClN₂O₂ |
MDL Number:
MFCD13562865 CAS Number: 1236261-42-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...